Gene Transcripts
- Gene Symbol
- Ptprcap
- Entrez Gene ID
- 19265
- Gene Name
- protein tyrosine phosphatase, receptor type, C polypeptide-associated protein
- Gene Aliases
- AI605264, CD-45-AP, LPAP, LSM-1
- Chromosome Location
- Chr. 19: 4154646 - 4156710 on Build GRCm38
- UniGene ID
- Mm.271937
- Species
- Mus musculus
- Gene Symbol
- Rps6kb2
- Entrez Gene ID
- 58988
- Gene Name
- ribosomal protein S6 kinase, polypeptide 2
- Gene Aliases
- 70kDa, S6K2
- Chromosome Location
- Chr. 19: 4156792 - 4163383 on Build GRCm38
- UniGene ID
- Mm.271937
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK090093.1 |
- |
|
2021 |
|
View Full Product Details