Gene Transcripts

Gene Symbol
Olfr70
Entrez Gene ID
56014
Gene Name
olfactory receptor 70
Gene Aliases
MOR262-10, mOR6
Chromosome Location
Chr. 4: 43695000 - 43700807 on Build GRCm38
UniGene ID
Mm.331571
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019485.2 NP_062358.2 2 607
GenBank mRNA BC046311.1 - 2 607

View Full Product Details