Gene Transcripts

Gene Symbol
Amtn
Entrez Gene ID
71421
Gene Name
amelotin
Gene Aliases
5430427O21Rik
Chromosome Location
Chr. 5: 88376108 - 88385916 on Build GRCm38
UniGene ID
Mm.79700
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_027793.1 NP_082069.1 8 521
GenBank mRNA AK017352.1 - 8 521

View Full Product Details