Gene Transcripts

Gene Symbol
Msl2
Entrez Gene ID
77853
Gene Name
male-specific lethal 2 homolog (Drosophila)
Gene Aliases
E130103E02Rik, Msl2l1, Rnf184
Chromosome Location
Chr. 9: 101074762 - 101075421 on Build GRCm38
UniGene ID
Mm.326206
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001100451.2 NP_001093921.1 1708
GenBank mRNA AK153696.1 - 2298
AK173208.1 - 2479
BC070432.1 - 1708
BC100371.1 - 1 862

View Full Product Details