Gene Transcripts

Gene Symbol
Ociad2
Entrez Gene ID
100361733
Gene Name
OCIA domain containing 2
Gene Aliases
Chromosome Location
Chr. 14: 37432999 - 37449425 on Build Rnor_6.0
UniGene ID
Rn.105660
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001271181.1 NP_001258110.1 4, 5 316
GenBank mRNA FM129783.1 - 5, 6 331
FM139638.1 - 4, 5 316
FQ214213.1 - 4, 5 305

View Full Product Details