Gene Transcripts

Gene Symbol
Slc17a9
Entrez Gene ID
362287
Gene Name
solute carrier family 17 (vesicular nucleotide transporter), member 9
Gene Aliases
RGD1311940
Chromosome Location
Chr. 3: 176228439 - 176246971 on Build Rnor_6.0
UniGene ID
Rn.101413
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001108613.1 NP_001102083.1 2 246
XM_006235769.2 XP_006235831.1 2 2185

View Full Product Details