Gene Transcripts

Gene Symbol
Olr45
Entrez Gene ID
293192
Gene Name
olfactory receptor 45
Gene Aliases
Chromosome Location
Chr. 1: 167748100 - 167749053 on Build Rnor_6.0
UniGene ID
Rn.141929
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001000124.1 NP_001000124.1 1 68

View Full Product Details