Gene Transcripts

Gene Symbol
Tab1
Entrez Gene ID
315139
Gene Name
TGF-beta activated kinase 1/MAP3K7 binding protein 1
Gene Aliases
Map3k7ip1
Chromosome Location
Chr. 7: 121350953 - 121382100 on Build Rnor_6.0
UniGene ID
Rn.17231
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001109976.2 NP_001103446.1 5 549
GenBank mRNA EV771639.1 - 3 320
FQ211519.1 - 5 984

View Full Product Details