Gene Transcripts

Gene Symbol
Tacstd2
Entrez Gene ID
494343
Gene Name
tumor-associated calcium signal transducer 2
Gene Aliases
Prp1
Chromosome Location
Chr. 4: 98341187 - 98342887 on Build Rnor_6.0
UniGene ID
Rn.24809
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001009540.2 NP_001009540.2 1 563
GenBank mRNA AY185508.1 - 1 563
BC060519.1 - 1 544 5597863

View Full Product Details