Gene Transcripts

Gene Symbol
Rmi2
Entrez Gene ID
497856
Gene Name
RecQ mediated genome instability 2
Gene Aliases
RGD1563163
Chromosome Location
Chr. 10: 4904458 - 4910339 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_573037.4 XP_573037.1 1 51

View Full Product Details