Gene Transcripts
- Gene Symbol
- Snora69
- Entrez Gene ID
- 104369
- Gene Name
- small nucleolar RNA, H/ACA box 69
- Gene Aliases
- MBI-134, Rnu69
- Chromosome Location
- Chr. X: 37082912 - 37083033 on Build GRCm38
- UniGene ID
- Mm.453531
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_002900.1 |
|
1 |
107 |
|
GenBank mRNA |
AF357420.1 |
- |
1 |
107 |
|
View Full Product Details