Gene Transcripts

Gene Symbol
PRKXP1
Entrez Gene ID
441733
Gene Name
protein kinase, X-linked, pseudogene 1
Gene Aliases
Chromosome Location
Chr. 15: 100547752 - 100559283 on Build GRCh38
UniGene ID
Hs.12250
Species
Homo sapiens
Interrogated Sequence Translated Protein RNADB ID Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_073405.1
5052
GenBank mRNA Y10483.1 - LIT2111
PRKXP1
2 242

View Full Product Details