Gene Transcripts
- Gene Symbol
- ZDHHC9
- Entrez Gene ID
- 51114
- Gene Name
- zinc finger DHHC-type containing 9
- Gene Aliases
- CGI89, CXorf11, DHHC9, MMSA1, MRXSZ, ZDHHC10, ZNF379, ZNF380
- Chromosome Location
- Chr. X: 129803288 - 129843934 on Build GRCh38
- UniGene ID
- Hs.193566
- Species
- Homo sapiens
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
AW969573.1 |
- |
1 |
228 |
|
Chr. X - 129803308 - 129803851 [-] |
GenBankmRNA |
DB543254.1 |
- |
1 |
174 |
|
Chr. X - 129803315 - 129803797 [-] |
View Full Product Details