Gene Transcripts

Gene Symbol
MAP3K8
Entrez Gene ID
1326
Gene Name
mitogen-activated protein kinase kinase kinase 8
Gene Aliases
AURA2, COT, EST, ESTF, MEKK8, TPL2, Tpl-2, c-COT
Chromosome Location
Chr. 10: 30434021 - 30461833 on Build GRCh38
UniGene ID
Hs.432453
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_017015709.1 XP_016871198.1 1 2092
GenBank mRNA BC022398.1 - 1 751

View Full Product Details