Gene Transcripts
- Gene Symbol
- Sfpq
- Entrez Gene ID
- 71514
- Gene Name
- splicing factor proline/glutamine rich (polypyrimidine tract binding protein associated)
- Gene Aliases
- 1110004P21Rik, 2810416M14Rik, 5730453G22Rik, 9030402K04Rik, AU021830, D4Ertd314e, Gm12940, OTTMUSG00000009329, PSF, REP1
- Chromosome Location
- Chr. 4: 127020560 - 127037016 on Build GRCm38
- UniGene ID
- Mm.257276
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK017658.1 |
- |
1 |
191 |
|
AK141302.1 |
- |
1 |
1485 |
|
BC033603.1 |
- |
1 |
2883 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BY717438.1 |
- |
1 |
191 |
|
Chr. 4 - 127034571 - 127034950 [+] |
GenBankmRNA |
BM934843.1 |
- |
1 |
320 |
|
Chr. 4 - 127034443 - 127035091 [+] |
GenBankmRNA |
CD770415.1 |
- |
1 |
154 |
|
Chr. 4 - 127034611 - 127035245 [+] |
GenBankmRNA |
CX207240.1 |
- |
1 |
277 |
|
Chr. 4 - 127034485 - 127034993 [+] |
View Full Product Details