Gene Transcripts

Gene Symbol
KLHL7-AS1
Entrez Gene ID
100775104
Gene Name
KLHL7 antisense RNA 1 (head to head)
Gene Aliases
PLATAK
Chromosome Location
Chr. 7: 23101228 - 23105703 on Build GRCh38
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_046220.1 3 2539
GenBank mRNA AK054880.1 - 3 2539

Gene Symbol
NFYC
Entrez Gene ID
4802
Gene Name
nuclear transcription factor Y subunit gamma
Gene Aliases
CBF-C, CBFC, H1TF2A, HAP5, HSM, NF-YC
Chromosome Location
Chr. 1: 40691570 - 40771603 on Build GRCh38
UniGene ID
Hs.713051
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_017001364.1 XP_016856853.1 1 208
XM_017001366.1 XP_016856855.1 1 142


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GENCODE ENST00000419813.1 - 3 2539 Chr. 7 - 23101228 - 23105703 [-]

View Full Product Details