Gene Transcripts
- Gene Symbol
- LINC00520
- Entrez Gene ID
- 645687
- Gene Name
- long intergenic non-protein coding RNA 520
- Gene Aliases
- C14orf34
- Chromosome Location
- Chr. 14: 55781135 - 55796674 on Build GRCh38
- UniGene ID
- Hs.569345
- Species
- Homo sapiens
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BU155797.1 |
- |
3 |
177 |
|
Chr. 14 - 55792742 - 55796639 [-] |
GenBankmRNA |
BQ232082.1 |
- |
3 |
252 |
|
Chr. 14 - 55782000 - 55796667 [-] |
View Full Product Details