Gene Transcripts
- Gene Symbol
- Rmi2
- Entrez Gene ID
- 223970
- Gene Name
- RMI2, RecQ mediated genome instability 2, homolog (S. cerevisiae)
- Gene Aliases
- A630055G03Rik, Gm537, Gm929
- Chromosome Location
- Chr. 16: 10835059 - 10843235 on Build GRCm38
- UniGene ID
- Mm.325777
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK080331.1 |
- |
|
413 |
|
View Full Product Details