Gene Transcripts

Gene Symbol
PMAIP1
Entrez Gene ID
5366
Gene Name
phorbol-12-myristate-13-acetate-induced protein 1
Gene Aliases
APR, NOXA
Chromosome Location
Chr. 18: 59899960 - 59904306 on Build GRCh38
UniGene ID
Hs.96
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_021127.2 NP_066950.1 2 1247
GenBank mRNA BC013120.1 - 2 1171
D90070.1 - 2 1201

View Full Product Details