Gene Transcripts

Gene Symbol
Wipf3
Entrez Gene ID
330319
Gene Name
WAS/WASL interacting protein family, member 3
Gene Aliases
9030621G03, BQ952480
Chromosome Location
Chr. 6: 54429218 - 54503768 on Build GRCm38
UniGene ID
Mm.434326
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001167860.1 NP_001161332.1 2, 3 257
NM_001167861.1 NP_001161333.1 2, 3 257

View Full Product Details