Gene Transcripts

Gene Symbol
Tmprss9
Entrez Gene ID
432478
Gene Name
transmembrane protease, serine 9
Gene Aliases
Serase-1B
Chromosome Location
Chr. 10: 80871117 - 80899495 on Build GRCm38
UniGene ID
Mm.235302
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001081688.2 NP_001075157.2 15 2727
GenBank mRNA BC158102.1 - 15 2823
BC172169.1 - 15 2823 9054839

View Full Product Details