Gene Transcripts

Gene Symbol
Aqp4
Entrez Gene ID
25293
Gene Name
aquaporin 4
Gene Aliases
AQP-4, MIWC2, Miwc
Chromosome Location
Chr. 18: 6766009 - 6782757 on Build Rnor_6.0
UniGene ID
Rn.90091
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001142366.2 NP_001135838.1 3, 4 691
NM_012825.4 NP_036957.1 3, 4 683
GenBank mRNA L27588.1 - 2, 3 551
U14007.1 - 3, 4 617

View Full Product Details