Gene Transcripts

Gene Symbol
Vax1
Entrez Gene ID
64571
Gene Name
ventral anterior homeobox 1
Gene Aliases
Chromosome Location
Chr. 1: 280334897 - 280338813 on Build Rnor_6.0
UniGene ID
Rn.48764
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_022636.1 NP_072158.1 2 413
GenBank mRNA AF113515.1 - 2 413

View Full Product Details