Gene Transcripts

Gene Symbol
Sval2
Entrez Gene ID
84605
Gene Name
seminal vesicle antigen-like 2
Gene Aliases
SLP-M
Chromosome Location
Chr. 4: 71340674 - 71344638 on Build Rnor_6.0
UniGene ID
Rn.74250
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_133291.1 NP_579825.1 2 112
GenBank mRNA AB051831.1 - 2 112

View Full Product Details