Gene Transcripts

Gene Symbol
Slc35e4
Entrez Gene ID
266687
Gene Name
solute carrier family 35, member E4
Gene Aliases
Chromosome Location
Chr. 14: 84163971 - 84170835 on Build Rnor_6.0
UniGene ID
Rn.15189
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_153316.2 NP_695228.2 2 1503
GenBank mRNA AF182714.1 - 3 1226
BC085693.1 - 2 1503 7191218

View Full Product Details