Gene Transcripts

Gene Symbol
Nubpl
Entrez Gene ID
299008
Gene Name
nucleotide binding protein-like
Gene Aliases
RGD1307232
Chromosome Location
Chr. 6: 72891758 - 73147837 on Build Rnor_6.0
UniGene ID
Rn.13455
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001185025.1 NP_001171954.1 7 586
XM_008764655.1 XP_008762877.1 7 541
GenBank mRNA CO399813.1 - 7 562
FQ210674.1 - 7 604

View Full Product Details