Gene Transcripts

Gene Symbol
Agfg2
Entrez Gene ID
304375
Gene Name
ArfGAP with FG repeats 2
Gene Aliases
Hrbl
Chromosome Location
Chr. 12: 22082041 - 22118239 on Build Rnor_6.0
UniGene ID
Rn.17775
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006249140.2 XP_006249202.1 4 739
XM_006249141.2 XP_006249203.1 4 738
XM_006249142.2 XP_006249204.1 4 740

View Full Product Details