Gene Transcripts

Gene Symbol
Mpp7
Entrez Gene ID
307035
Gene Name
membrane protein, palmitoylated 7 (MAGUK p55 subfamily member 7)
Gene Aliases
Chromosome Location
Chr. 17: 60057085 - 60250303 on Build Rnor_6.0
UniGene ID
Rn.21436
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001100575.1 NP_001094045.1 5 505
XM_006254138.2 XP_006254200.1 6 2288
GenBank mRNA BC085813.1 - 5 588

View Full Product Details