Gene Transcripts

Gene Symbol
Samd7
Entrez Gene ID
310257
Gene Name
sterile alpha motif domain containing 7
Gene Aliases
Chromosome Location
Chr. 2: 116243243 - 116262612 on Build Rnor_6.0
UniGene ID
Rn.225098
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001191703.1 NP_001178632.1 2 121
XM_006232225.2 XP_006232287.1 3 645

View Full Product Details