Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001139517.1 | NP_001132989.1 | 6 | 984 | |
NM_001139518.1 | NP_001132990.1 | 7 | 705 | ||
NM_001316362.1 | NP_001303291.1 | 8 | 1023 | ||
NM_003690.4 | NP_003681.1 | 7 | 904 | ||
XM_011512063.1 | XP_011510365.1 | 7 | 835 | ||
XM_011512066.1 | XP_011510368.1 | 8 | 1060 | ||
XM_017005159.1 | XP_016860648.1 | 8 | 945 | ||
GenBank mRNA | AF072860.1 | - | 7 | 799 | |
AF083033.1 | - | 7 | 795 | ||
AK093360.1 | - | 3 | 930 | ||
AK223107.1 | - | 7 | 803 | ||
AK290601.1 | - | 7 | 830 | ||
AK296019.1 | - | 8 | 934 | ||
AL833867.1 | - | 6 | 984 | ||
AY251164.1 | - | 7 | 722 | ||
BC009470.1 | - | 7 | 810 | 3503747 | |
BT007243.1 | - | 7 | 703 |