Gene Transcripts

Gene Symbol
Ttc39d
Entrez Gene ID
366531
Gene Name
tetratricopeptide repeat domain 39D
Gene Aliases
RGD1311615
Chromosome Location
Chr. 6: 2886764 - 2895118 on Build Rnor_6.0
UniGene ID
Rn.103605
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001191906.1 NP_001178835.1 2 689

View Full Product Details