Gene Transcripts

Gene Symbol
Samd10
Entrez Gene ID
499957
Gene Name
sterile alpha motif domain containing 10
Gene Aliases
RGD1563203
Chromosome Location
Chr. 3: 177091467 - 177095598 on Build Rnor_6.0
UniGene ID
Rn.139640
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001191967.1 NP_001178896.1 3, 4 456
XM_006235798.1 XP_006235860.1 3, 4 467

View Full Product Details