Gene Transcripts
- Gene Symbol
- Catsper4
- Entrez Gene ID
- 362623
- Gene Name
- cation channel, sperm associated 4
- Gene Aliases
- RGD1565213
- Chromosome Location
- Chr. 5: 152430048 - 152446808 on Build Rnor_6.0
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
XM_342941.7 |
XP_342942.5
|
8, 9 |
1041 |
|
View Full Product Details