Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001005417.2 | NP_001005417.1 | 6 | 1106 | |
NM_003780.4 | NP_003771.1 | 6 | 1306 | ||
NM_030587.2 | NP_085076.2 | 6 | 1107 | ||
XM_017002716.1 | XP_016858205.1 | 6 | 1064 | ||
XM_017002717.1 | XP_016858206.1 | 6 | 1267 | ||
GenBank mRNA | AB024434.1 | - | 5 | 952 | |
AK074533.1 | - | 4 | 604 | ||
AK095873.1 | - | 6 | 1734 | ||
AK293418.1 | - | 6 | 1056 | ||
AL137647.1 | - | 4 | 501 | ||
BC096821.1 | - | 6 | 1074 | 30558762 | |
Y12510.1 | - | 5 | 944 |