Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_029760.2 | NP_084036.2 | 9 | 834 | |
GenBank mRNA | AK010827.1 | - | 9 | 816 | |
AK141257.1 | - | 9 | 798 | ||
AK152377.1 | - | 9 | 835 | ||
BC023294.1 | - | 428 | |||
BC062175.1 | - | 9 | 815 | 30291107 |