Gene Transcripts

Gene Symbol
Sbpl
Entrez Gene ID
638345
Gene Name
spermine binding protein-like
Gene Aliases
2310068J22Rik
Chromosome Location
Chr. 17: 23953085 - 23955273 on Build GRCm38
UniGene ID
Mm.46428
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001077421.1 NP_001070889.1 3 506
GenBank mRNA AK009270.1 - 3 508
AK009356.1 - 3 496
AK009376.1 - 3 507
AK009398.1 - 3 508
AK009406.1 - 3 507
AK009446.1 - 3 507
AK009467.1 - 3 507
AK009489.1 - 3 507
AK009501.1 - 3 505
AK009509.1 - 3 507
AK009539.1 - 3 507
AK009553.1 - 3 506
AK009598.1 - 3 507
AK009609.1 - 3 507
AK009613.1 - 3 507
AK009627.1 - 3 509
AK009630.1 - 3 506
AK009631.1 - 3 510
AK009647.1 - 3 505
AK009712.1 - 3 507
AK009758.1 - 3 505
AK009761.1 - 3 505
AK009792.1 - 3 505
AK009819.1 - 3 507
AK009834.1 - 3 507
AK009845.1 - 3 507
AK009910.1 - 3 506
AK009917.1 - 3 508
AK009931.1 - 3 509
AK009982.1 - 3 506
AK010076.1 - 3 506
AK010078.1 - 3 507
AK010083.1 - 3 507
AK010102.1 - 3 507
AK010114.1 - 3 507
AK010161.1 - 3 507
AK010189.1 - 3 507
AK010216.1 - 3 507
AK010221.1 - 3 506
AK010237.1 - 3 506
AK075878.1 - 2 386
AK075905.1 - 1 235
AK160518.1 - 3 506
BC089471.1 - 3 504 30301294

View Full Product Details