Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_012435.2 | NP_036567.2 | 10, 11 | 1301 | |
XM_011527893.2 | XP_011526195.1 | 10, 11 | 1316 | ||
XM_011527896.2 | XP_011526198.1 | 12, 13 | 1361 | ||
XM_017026567.1 | XP_016882056.1 | 11, 12 | 1449 | ||
GenBank mRNA | AB001451.1 | - | 10, 11 | 1175 | |
AL360254.1 | - | 8, 9 | 731 | ||
BY796046.2 | - | 1, 2 | 118 |