Gene Transcripts
- Gene Symbol
- PHF19
- Entrez Gene ID
- 26147
- Gene Name
- PHD finger protein 19
- Gene Aliases
- MTF2L1, PCL3, TDRD19B
- Chromosome Location
- Chr. 9: 120855651 - 120901967 on Build GRCh38
- UniGene ID
- Hs.460124
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001009936.2 |
NP_001009936.1
|
2, 3 |
447 |
|
NM_001286840.1 |
NP_001273769.1
|
2, 3 |
318 |
|
NM_001286843.1 |
NP_001273772.1
|
2, 3 |
447 |
|
NM_015651.2 |
NP_056466.1
|
2, 3 |
447 |
|
XM_005251906.2 |
XP_005251963.1
|
2, 3 |
255 |
|
XM_011518509.2 |
XP_011516811.1
|
2, 3 |
298 |
|
XM_011518511.2 |
XP_011516813.1
|
2, 3 |
781 |
|
XM_011518515.2 |
XP_011516817.1
|
2, 3 |
782 |
|
XM_011518516.2 |
XP_011516818.1
|
2, 3 |
784 |
|
XM_017014612.1 |
XP_016870101.1
|
2, 3 |
287 |
|
XM_017014613.1 |
XP_016870102.1
|
2, 3 |
785 |
|
GenBank mRNA |
AK122744.1 |
- |
2, 3 |
307 |
|
BC022374.1 |
- |
2, 3 |
307 |
|
BC108663.1 |
- |
2, 3 |
414 |
|
BC125076.1 |
- |
1, 2 |
219 |
40117657 |
BC125077.1 |
- |
1, 2 |
219 |
40117658 |
BM784783.1 |
- |
2, 3 |
447 |
|
BX640713.1 |
- |
2, 3 |
319 |
|
View Full Product Details