Gene Transcripts

Gene Symbol
TMEM55A
Entrez Gene ID
55529
Gene Name
transmembrane protein 55A
Gene Aliases
Chromosome Location
Chr. 8: 90994271 - 91040975 on Build GRCh38
UniGene ID
Hs.202517
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_018710.2 NP_061180.1 7 915
GenBank mRNA AK313783.1 - 7 828
AX775961.1 - 7 765
BC033892.1 - 7 765 4820065
CR749733.1 - 6 479

View Full Product Details