Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_020645.2 | NP_065696.1 | 5 | 700 | |
GenBank mRNA | AK024750.1 | - | 5 | 653 | |
AK098694.1 | - | 5 | 653 | ||
AK296484.1 | - | 4 | 512 | ||
AY211930.1 | - | 4 | 397 | ||
BC008835.2 | - | 5 | 636 | 4304488 |