Gene Transcripts
- Gene Symbol
- SLC17A9
- Entrez Gene ID
- 63910
- Gene Name
- solute carrier family 17 member 9
- Gene Aliases
- C20orf59, POROK8, VNUT
- Chromosome Location
- Chr. 20: 62952647 - 62968597 on Build GRCh38
- UniGene ID
- Hs.512686
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001302643.1 |
NP_001289572.1
|
12, 13 |
1357 |
|
NM_022082.3 |
NP_071365.3
|
11, 12 |
1295 |
|
XM_011528978.2 |
XP_011527280.1
|
11, 12 |
1207 |
|
GenBank mRNA |
AK027065.1 |
- |
4, 5 |
284 |
|
AK074107.1 |
- |
10, 11 |
2091 |
|
AK095473.1 |
- |
11, 12 |
1146 |
|
AK125597.1 |
- |
11, 12 |
1818 |
|
AK298503.1 |
- |
11, 12 |
1201 |
|
BC025312.1 |
- |
12, 13 |
1297 |
4563089 |
BC027447.1 |
- |
7, 8 |
516 |
|
BC038593.1 |
- |
11, 12 |
1137 |
|
View Full Product Details