Gene Transcripts
- Gene Symbol
- NUBPL
- Entrez Gene ID
- 80224
- Gene Name
- nucleotide binding protein like
- Gene Aliases
- C14orf127, IND1, huInd1
- Chromosome Location
- Chr. 14: 31561385 - 31861293 on Build GRCh38
- UniGene ID
- Hs.288981
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001201573.1 |
NP_001188502.1
|
2, 3 |
200 |
|
NM_025152.2 |
NP_079428.2
|
4, 5 |
436 |
|
NR_120408.1 |
|
4, 5 |
436 |
|
XM_011537183.2 |
XP_011535485.1
|
4, 5 |
422 |
|
XM_017021664.1 |
XP_016877153.1
|
4, 5 |
422 |
|
XM_017021665.1 |
XP_016877154.1
|
4, 5 |
422 |
|
XM_017021666.1 |
XP_016877155.1
|
4, 5 |
422 |
|
GenBank mRNA |
AK022722.1 |
- |
4, 5 |
349 |
|
AK295326.1 |
- |
4, 5 |
397 |
|
AK301447.1 |
- |
2, 3 |
200 |
|
AK316445.1 |
- |
4, 5 |
426 |
|
BC024919.1 |
- |
4, 5 |
362 |
4475196 |
BX248028.1 |
- |
4, 5 |
423 |
|
KJ534907.1 |
- |
4, 5 |
381 |
|
KJ534908.1 |
- |
4, 5 |
381 |
|
KJ534909.1 |
- |
4, 5 |
381 |
|
View Full Product Details