Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001127496.1 | NP_001120968.1 | 2 | 408 | |
NM_001293289.1 | NP_001280218.1 | 3 | 786 | ||
NM_001293290.1 | NP_001280219.1 | 2 | 724 | ||
NM_030964.3 | NP_112226.2 | 3 | 459 | ||
XM_011537685.2 | XP_011535987.1 | 3 | 1186 | ||
XM_017009910.1 | XP_016865399.1 | 2 | 1452 | ||
GenBank mRNA | AF227516.1 | - | 3 | 405 | |
AK096464.1 | - | 2 | 393 | ||
AK226147.1 | - | 1 | 831 | ||
BC125095.1 | - | 2 | 257 | 40118267 | |
BC125096.1 | - | 2 | 257 | 40118268 | |
DA183391.1 | - | 2 | 408 |