Gene Transcripts

Gene Symbol
C15orf41
Entrez Gene ID
84529
Gene Name
chromosome 15 open reading frame 41
Gene Aliases
HH114
Chromosome Location
Chr. 15: 36579603 - 36810260 on Build GRCh38
UniGene ID
Hs.48348
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001130010.2 NP_001123482.1 10 896
NM_001290232.1 NP_001277161.1 10 587
NM_001290233.1 NP_001277162.1 10 896
NM_001321756.1 NP_001308685.1 9 850
NM_001321757.1 NP_001308686.1 11 930
NM_001321758.1 NP_001308687.1 8 785
NM_001321759.1 NP_001308688.1 10 896
NM_001321761.1 NP_001308690.1 10 914
NM_032499.5 NP_115888.1 11 642
XM_011522111.1 XP_011520413.1 10 888
XM_017022676.1 XP_016878165.1 7 690
XM_017022677.1 XP_016878166.1 7 690
GenBank mRNA AF114263.1 - 11 617
AK026504.1 - 10 865
AK315446.1 - 10 556
BC006254.2 - 10 655 3952242
BC042750.1 - 2 5001
BX647650.1 - 4 266

View Full Product Details