Gene Transcripts

Gene Symbol
TTC6
Entrez Gene ID
319089
Gene Name
tetratricopeptide repeat domain 6
Gene Aliases
C14orf25, NCRNA00291
Chromosome Location
Chr. 14: 37595199 - 37842628 on Build GRCh38
UniGene ID
Hs.659936
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001310135.1 NP_001297064.1 33 6451
XM_011537430.2 XP_011535732.1 29 4283
XM_011537431.2 XP_011535733.1 27 3940
XM_017021254.1 XP_016876743.1 32 6301
XM_017021255.1 XP_016876744.1 31 6166
XM_017021256.1 XP_016876745.1 32 6565
GenBank mRNA BC014342.2 - 6 858
BC103914.2 - 15 2021 40006180
BC103915.1 - 14 1800 40006182
BC103916.2 - 14 1800 40006183
BX161415.1 - 13 1735

View Full Product Details