Gene Transcripts
- Gene Symbol
- TTC6
- Entrez Gene ID
- 319089
- Gene Name
- tetratricopeptide repeat domain 6
- Gene Aliases
- C14orf25, NCRNA00291
- Chromosome Location
- Chr. 14: 37595199 - 37842628 on Build GRCh38
- UniGene ID
- Hs.659936
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001310135.1 |
NP_001297064.1
|
30, 31 |
6037 |
|
XM_011537430.2 |
XP_011535732.1
|
26, 27 |
3869 |
|
XM_011537431.2 |
XP_011535733.1
|
24, 25 |
3526 |
|
XM_017021254.1 |
XP_016876743.1
|
29, 30 |
5887 |
|
XM_017021255.1 |
XP_016876744.1
|
28, 29 |
5752 |
|
XM_017021256.1 |
XP_016876745.1
|
29, 30 |
6151 |
|
GenBank mRNA |
BC014342.2 |
- |
3, 4 |
444 |
|
BC103914.2 |
- |
11, 12 |
1386 |
40006180 |
BC103915.1 |
- |
11, 12 |
1386 |
40006182 |
BC103916.2 |
- |
11, 12 |
1386 |
40006183 |
BX161415.1 |
- |
10, 11 |
1321 |
|
View Full Product Details