Gene Transcripts

Gene Symbol
NLRP5
Entrez Gene ID
126206
Gene Name
NLR family pyrin domain containing 5
Gene Aliases
CLR19.8, MATER, NALP5, PAN11, PYPAF8
Chromosome Location
Chr. 19: 55999726 - 56061810 on Build GRCh38
UniGene ID
Hs.356872
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_153447.4 NP_703148.4 7 2140
GenBank mRNA AY054986.1 - 7 2140
AY154460.1 - 7 2140

View Full Product Details