Gene Transcripts

Gene Symbol
OCIAD2
Entrez Gene ID
132299
Gene Name
OCIA domain containing 2
Gene Aliases
Chromosome Location
Chr. 4: 48885380 - 48906828 on Build GRCh38
UniGene ID
Hs.95835
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001014446.2 NP_001014446.1 2, 3 309
NM_001286773.1 NP_001273702.1 2, 3 332
NM_152398.3 NP_689611.1 2, 3 309
GenBank mRNA AK298305.1 - 2, 3 278
AY472117.1 - 2, 3 311
BC032808.1 - 2, 3 280 4454708
BI546814.1 - 2, 3 314
BM782144.1 - 2, 3 279
CR749310.1 - 2, 3 254
W68799.1 - 2, 3 261

View Full Product Details