Gene Transcripts
- Gene Symbol
- C3orf22
- Entrez Gene ID
- 152065
- Gene Name
- chromosome 3 open reading frame 22
- Gene Aliases
- Chromosome Location
- Chr. 3: 126524439 - 126558965 on Build GRCh38
- UniGene ID
- Hs.178210
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_152533.2 |
NP_689746.1
|
2, 3 |
461 |
|
NR_130715.1 |
|
2, 3 |
461 |
|
XM_011512455.2 |
XP_011510757.1
|
2, 3 |
461 |
|
XM_011512457.2 |
XP_011510759.1
|
2, 3 |
461 |
|
XM_017005751.1 |
XP_016861240.1
|
2, 3 |
431 |
|
XM_017005752.1 |
XP_016861241.1
|
2, 3 |
293 |
|
XM_017005753.1 |
XP_016861242.1
|
2, 3 |
461 |
|
GenBank mRNA |
AK097841.1 |
- |
2, 3 |
461 |
|
BC032025.1 |
- |
2, 3 |
411 |
5165128 |
DB055549.1 |
- |
2, 3 |
461 |
|
View Full Product Details