Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_174908.3 | NP_777568.1 | 6 | 1111 | |
NM_178335.2 | NP_848018.1 | 7 | 1639 | ||
GenBank mRNA | AJ416916.2 | - | 6 | 1111 | |
AJ557013.1 | - | 7 | 1639 | ||
BC065004.1 | - | 6 | 608 | 6049272 |